accessibility menu, dialog, popup

UserName

CH3NH3CSNCH3H3C

CAS RN 2782-91-4

IUPAC Name: 1,1,3,3-tetramethylthiourea
Synonyms: tetramethylthiourea
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12N2S
InChi Key: MNOILHPDHOHILI-UHFFFAOYSA-N
SMILES: CN(C)C(=S)N(C)C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results