accessibility menu, dialog, popup

UserName

CAS RN 27848-80-2

IUPAC Name: lithium(1+) 2-hydroxypropanoate
Synonyms: lithium(1+) lactate
Molecular Weight (g/mol): 96.01
Molecular Formula: C3H5LiO3
InChi Key: GKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES: [Li+].CC(O)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (2)
  • (4)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)