accessibility menu, dialog, popup

UserName

CAS RN 28097-03-2

CH3NONHNOOHSSSSHONH3COONHN

CAS RN 28097-03-2

IUPAC Name: 14-(hydroxymethyl)-3-[14-(hydroxymethyl)-18-methyl-13,17-dioxo-15,16-dithia-10,12,18-triazapentacyclo[12.2.2.0¹,¹².0³,¹¹.0⁴,⁹]octadeca-4,6,8-trien-3-yl]-18-methyl-15,16-dithia-10,12,18-triazapentacyclo[12.2.2.0¹,¹².0³,¹¹.0⁴,⁹]octadeca-4,6,8-triene-13,17-dione
Synonyms: chaetocin
Molecular Weight (g/mol): 696.83
Molecular Formula: C30H28N6O6S4
InChi Key: PZPPOCZWRGNKIR-UHFFFAOYNA-N
SMILES: CN1C(=O)C23CC4(C(NC5=CC=CC=C45)N2C(=O)C1(CO)SS3)C12CC34SSC(CO)(N(C)C3=O)C(=O)N4C1NC1=CC=CC=C21
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)