accessibility menu, dialog, popup

UserName

CAS RN 28240-69-9

ClPClPClCl

CAS RN 28240-69-9

IUPAC Name: dichloro[2-(dichlorophosphanyl)ethyl]phosphane
Synonyms: dichloro[2-(dichlorophosphanyl)ethyl]phosphane
Molecular Weight (g/mol): 231.8
Molecular Formula: C2H4Cl4P2
InChi Key: SBWAJHLQMFBNIN-UHFFFAOYSA-N
SMILES: ClP(Cl)CCP(Cl)Cl
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)