accessibility menu, dialog, popup

UserName

COCOCOCOCOCOCOCOCOCOCOCOCOCOCOCORhRhRhRhRhRh

CAS RN 28407-51-4

IUPAC Name: hexadecakis(methanidylidyneoxidanium) hexarhodium
Synonyms: hexadecacarbonylhexarhodium
Molecular Weight (g/mol): 1065.59
Molecular Formula: C16O16Rh6
InChi Key: SZQABOJVTZVBHE-UHFFFAOYSA-N
SMILES: [Rh].[Rh].[Rh].[Rh].[Rh].[Rh].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results