accessibility menu, dialog, popup

UserName

CAS RN 2855-13-2

H2NH2NCH3CH3CH3

CAS RN 2855-13-2

IUPAC Name: 3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine; 4-methylbenzene-1-sulfonic acid
Synonyms: isophorone diamine; para-toluene sulfonate
Molecular Weight (g/mol): 342.5
Molecular Formula: C17H30N2O3S
InChi Key: ZBVBXDSQFYHMPD-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC1(C)CC(N)CC(C)(CN)C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)