accessibility menu, dialog, popup

UserName

ClH3CNCH3NNNNNCH3CH3(E/Z)

CAS RN 2869-83-2

IUPAC Name: 3-(diethylamino)-7-{2-[4-(dimethylamino)phenyl]diazen-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: janus green B
Molecular Weight (g/mol): 511.07
Molecular Formula: C30H31ClN6
InChi Key: XXACTDWGHQXLGW-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)