accessibility menu, dialog, popup

UserName

CAS RN 28697-17-8

IUPAC Name: 1-[(tert-butoxy)carbonyl]piperidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)piperidine-2-carboxylic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: JQAOHGMPAAWWQO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)