CAS RN 287917-96-8
4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 287917-96-8 Molecular Formula: C5H5BrN2O Molecular Weight (g/mol): 189.012 MDL Number: MFCD00103235 InChI Key: KGIYMMPLYIITLL-UHFFFAOYSA-N PubChem CID: 2735595 IUPAC Name: 4-bromo-1-methylpyrazole-3-carbaldehyde SMILES: CN1C=C(C(=N1)C=O)Br
4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 287917-96-8 Molecular Formula: C5H5BrN2O Molecular Weight (g/mol): 189.01 InChI Key: KGIYMMPLYIITLL-UHFFFAOYSA-N Synonym: 4-bromo-1-methyl-1h-pyrazole-3-carbaldehyde,4-bromo-3-formyl-1-methyl-1h-pyrazole,4-bromo-1-methylpyrazole-3-carboxaldehyde,4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde,1h-pyrazole-3-carboxaldehyde, 4-bromo-1-methyl,pubchem23668,acmc-209h4p,buttpark 88\08-06,4-bromo-3-formyl-1-methylpyrazole,1h-pyrazole-3-carboxaldehyde,4-bromo-1-methyl PubChem CID: 2735595 IUPAC Name: 4-bromo-1-methylpyrazole-3-carbaldehyde SMILES: CN1C=C(C(=N1)C=O)Br