accessibility menu, dialog, popup

UserName

OHOHOO

CAS RN 2892-51-5

IUPAC Name: dihydroxycyclobut-3-ene-1,2-dione
Synonyms: squaric acid quadratsaeure quadratic acid
Molecular Weight (g/mol): 114.06
Molecular Formula: C4H2O4
InChi Key: PWEBUXCTKOWPCW-UHFFFAOYSA-N
SMILES: OC1=C(O)C(=O)C1=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)