accessibility menu, dialog, popup

UserName

CAS RN 28954-12-3

H3C(S)OH(S)NH2OHO

CAS RN 28954-12-3

IUPAC Name: 2-amino-3-hydroxybutanoic acid
Synonyms: threonine(L) DL-allothreonine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: AYFVYJQAPQTCCC-UHFFFAOYNA-N
SMILES: CC(O)C(N)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)