CAS RN 2919946-71-5
CAS RN 2919946-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-Salbutamol, TRC
CAS: 2919946-71-5 Molecular Formula: C13 H20 N2 O4 Molecular Weight (g/mol): 268.31 Synonym: N-(tert-Butyl)-N-(2-hydroxy-2-(4-hydroxy-3-(hydroxymethyl)phenyl)ethyl)nitrous Amide,N-Nitrososalbutamol,N-Tert-butyl-N-[2-hydroxy-2-[4-hydroxy-3-(hydroxymethyl)phenyl]ethyl]nitrous amide,N-Nitrososalbutamol IUPAC Name: N-tert-butyl-N-[2-hydroxy-2-[4-hydroxy-3-(hydroxymethyl)phenyl]ethyl]nitrous amide SMILES: CC(C)(C)N(CC(O)c1ccc(O)c(CO)c1)N=O