accessibility menu, dialog, popup

UserName

CH3OOCH3BrOOH3C

CAS RN 29263-94-3

IUPAC Name: 1,3-diethyl 2-bromo-2-methylpropanedioate
Synonyms: 1,3-diethyl 2-bromo-2-methylpropanedioate
Molecular Weight (g/mol): 253.09
Molecular Formula: C8H13BrO4
InChi Key: CSLQAXTUGPUBCW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(Br)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results