accessibility menu, dialog, popup

UserName

BrHSnHBr

CAS RN 2954-94-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 446.84
Molecular Formula: C12H24Br2Sn
InChi Key: DSLZARZWEQQPGE-UHFFFAOYSA-N
SMILES: Br.Br.C1CCC(CC1)[SnH2]C1CCCCC1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results