accessibility menu, dialog, popup

UserName

CAS RN 29602-39-9

H3NNHHNNOO

CAS RN 29602-39-9

IUPAC Name: 2-[(2-azaniumylethyl)amino]-5-nitropyridin-1-ium
Synonyms: 2-[(2-ammonioethyl)amino]-5-nitropyridin-1-ium
Molecular Weight (g/mol): 184.2
Molecular Formula: C7H12N4O2
InChi Key: ODHSPTHLPCXPTL-UHFFFAOYSA-P
SMILES: [NH3+]CCNC1=CC=C(C=[NH+]1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results