accessibility menu, dialog, popup

UserName

OOOO

CAS RN 29745-04-8

IUPAC Name: 4,10-dioxatricyclo[5.2.1.0²,⁶]decane-3,5-dione
Synonyms: norcantharidin
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: JAABVEXCGCXWRR-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C3CCC(O3)C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)