CAS RN 2985926-49-4
CAS RN 2985926-49-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methapyrilene-d6 Hydrochloride, TRC
CAS: 2985926-49-4 Molecular Formula: C14H13D6N3S . HCl Molecular Weight (g/mol): 303.88 Synonym: 2-[[2-(Dimethylamino)ethyl]-2-thenylamino]pyridine-d6 hydrochloride,Thenylene-d6 hydrochloride,Thenylpyramine-d6 hydrochloride IUPAC Name: N1,N1-bis(methyl-d3)-N2-(pyridin-2-yl)-N2-(thiophen-2-ylmethyl)ethane-1,2-diamine hydrochloride SMILES: [2H]C([2H])([2H])N(C([2H])([2H])[2H])CCN(CC1=CC=CS1)C2=NC=CC=C2.Cl