accessibility menu, dialog, popup

UserName

CH3CH3CH3O

CAS RN 2987-16-8

IUPAC Name: 3,3-dimethylbutanal
Synonyms: 3,3-dimethylbutanal
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: LTNUSYNQZJZUSY-UHFFFAOYSA-N
SMILES: CC(C)(C)CC=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)