accessibility menu, dialog, popup

UserName

CAS RN 29881-14-9

IUPAC Name: (2-phenylcyclopropyl)benzene
Synonyms: (2-phenylcyclopropyl)benzene
Molecular Weight (g/mol): 194.28
Molecular Formula: C15H14
InChi Key: ZSIYTDQNAOYUNE-UHFFFAOYNA-N
SMILES: C1C(C1C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)