accessibility menu, dialog, popup

UserName

CAS RN 300557-77-1

IUPAC Name: 11-phenyl-10-sulfanylidene-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6)-dien-12-one
Synonyms: 11-phenyl-10-sulfanylidene-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6)-dien-12-one
Molecular Weight (g/mol): 300.39
Molecular Formula: C15H12N2OS2
InChi Key: LLFBYHAEKFMVIS-UHFFFAOYSA-N
SMILES: O=C1N(C(=S)NC2=C1C1=C(CCC1)S2)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results