accessibility menu, dialog, popup

UserName

NO

CAS RN 30078-65-0

IUPAC Name: furan-3-carbonitrile
Synonyms: furan-3-carbonitrile
Molecular Weight (g/mol): 93.09
Molecular Formula: C5H3NO
InChi Key: ZQFYJHMUAWCEBH-UHFFFAOYSA-N
SMILES: N#CC1=COC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results