accessibility menu, dialog, popup

UserName

CH3NOCH3SOCH3(E/Z)

CAS RN 300801-52-9

IUPAC Name: 1-(3-ethyl-5-methoxy-2,3-dihydro-1,3-benzothiazol-2-ylidene)propan-2-one
Synonyms: 1-(3-ethyl-5-methoxy-1,3-benzothiazol-2-ylidene)propan-2-one
Molecular Weight (g/mol): 249.33
Molecular Formula: C13H15NO2S
InChi Key: BGVLELSCIHASRV-UHFFFAOYSA-N
SMILES: CCN1C(SC2=C1C=C(OC)C=C2)=CC(C)=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results