accessibility menu, dialog, popup

UserName

CAS RN 30091-54-4

CH3NNNNCH3

CAS RN 30091-54-4

IUPAC Name: 5,6-dimethyl-3-(pyridin-2-yl)-1,2,4-triazine
Synonyms: 5,6-dimethyl-3-(pyridin-2-yl)-1,2,4-triazine
Molecular Weight (g/mol): 186.22
Molecular Formula: C10H10N4
InChi Key: NAFPPMVYCAHBAY-UHFFFAOYSA-N
SMILES: CC1=C(C)N=C(N=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results