accessibility menu, dialog, popup

UserName

CAS RN 301-02-0

IUPAC Name: (9E)-octadec-9-enamide
Synonyms: elaidoylamide
Molecular Weight (g/mol): 281.48
Molecular Formula: C18H35NO
InChi Key: FATBGEAMYMYZAF-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results