accessibility menu, dialog, popup

UserName

CAS RN 302841-89-0

H3COONHOO

CAS RN 302841-89-0

IUPAC Name: butyl N-(9H-xanthene-9-carbonyl)carbamate
Synonyms: butyl N-(9H-xanthene-9-carbonyl)carbamate
Molecular Weight (g/mol): 325.36
Molecular Formula: C19H19NO4
InChi Key: RQBUXEUMZZQUFY-UHFFFAOYSA-N
SMILES: CCCCOC(=O)NC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results