accessibility menu, dialog, popup

UserName

CAS RN 30389-18-5

H2NCH

CAS RN 30389-18-5

IUPAC Name: 1-ethynylcyclohexan-1-amine
Synonyms: 1-ethynylcyclohexan-1-amine
Molecular Weight (g/mol): 123.2
Molecular Formula: C8H13N
InChi Key: GDKOYYDQISQOMH-UHFFFAOYSA-N
SMILES: NC1(CCCCC1)C#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results