accessibility menu, dialog, popup

UserName

H3COBrF

CAS RN 304445-49-6

IUPAC Name: 1-(4-bromo-3-fluorophenyl)ethan-1-one
Synonyms: 1-(4-bromo-3-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: VCTWSAITPPCBHI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results