accessibility menu, dialog, popup

UserName

H2NOHN(S)HNNOHO

CAS RN 305-84-0

IUPAC Name: (2S)-2-(3-aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid
Synonyms: β-alanyl-L-histidine
Molecular Weight (g/mol): 226.24
Molecular Formula: C9H14N4O3
InChi Key: CQOVPNPJLQNMDC-ZETCQYMHSA-N
SMILES: NCCC(=O)N[C@@H](CC1=CN=CN1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)