accessibility menu, dialog, popup

UserName

H3CNOOHNN

CAS RN 305381-67-3

IUPAC Name: 1-methyl-1H-1,2,3-benzotriazole-5-carboxylic acid
Synonyms: 1-methyl-1,2,3-benzotriazole-5-carboxylic acid
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: SGHWYTLJLHVIBQ-UHFFFAOYSA-N
SMILES: CN1N=NC2=CC(=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results