accessibility menu, dialog, popup

UserName

CAS RN 305811-38-5

CH3NSH2N

CAS RN 305811-38-5

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)aniline
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)aniline
Molecular Weight (g/mol): 190.26
Molecular Formula: C10H10N2S
InChi Key: AJFBHXXQTQAILP-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results