accessibility menu, dialog, popup

UserName

CAS RN 306934-78-1

IUPAC Name: 4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine
Synonyms: 4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine
Molecular Weight (g/mol): 260.74
Molecular Formula: C13H9ClN2S
InChi Key: LRTKDTZXCFGPEU-UHFFFAOYSA-N
SMILES: CC1=C(SC2=NC=NC(Cl)=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results