accessibility menu, dialog, popup

UserName

CH3NONH2

CAS RN 306935-01-3

IUPAC Name: 1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
Synonyms: 1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: KJGJWCJXSLAKKS-UHFFFAOYSA-N
SMILES: CC1=C(CN)C(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results