accessibility menu, dialog, popup

UserName

CH3OFFFNCO

CAS RN 306935-03-5

IUPAC Name: 3-isocyanato-5-methyl-2-(trifluoromethyl)furan
Synonyms: 3-isocyanato-5-methyl-2-(trifluoromethyl)furan
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: WTKWFNIIIXNTDO-UHFFFAOYSA-N
SMILES: CC1=CC(N=C=O)=C(O1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results