accessibility menu, dialog, popup

UserName

CAS RN 306935-33-1

H3CH3CCH3OOOCH3Cl

CAS RN 306935-33-1

IUPAC Name: methyl 2-chloro-4,4-dimethyl-3-oxopentanoate
Synonyms: methyl 2-chloro-4,4-dimethyl-3-oxopentanoate
Molecular Weight (g/mol): 192.64
Molecular Formula: C8H13ClO3
InChi Key: NGRPVOKPBYTXLT-UHFFFAOYNA-N
SMILES: COC(=O)C(Cl)C(=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results