accessibility menu, dialog, popup

UserName

NNOOH

CAS RN 307307-97-7

IUPAC Name: 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxylic acid
Synonyms: 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: VOAKNFVZEGNOKV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2CCCCN2N=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)