accessibility menu, dialog, popup

UserName

H3C(S)(S)NHOHOCH3H3CO

CAS RN 3077-46-1

IUPAC Name: 2-acetamido-3-methylpentanoic acid
Synonyms: D-alloisoleucine, N-acetyl- DL-alloisoleucine, N-acetyl- L-alloisoleucine, N-acetyl-
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: JDTWZSUNGHMMJM-UHFFFAOYNA-N
SMILES: CCC(C)C(NC(C)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)