accessibility menu, dialog, popup

UserName

CH3OHOOH3C

CAS RN 30934-97-5

IUPAC Name: 2,2-dimethoxyethan-1-ol
Synonyms: 2,2-dimethoxyethanol
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: NYPNCQTUZYWFGG-UHFFFAOYSA-N
SMILES: COC(CO)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results