CAS RN 30981-98-7
CAS RN 30981-98-7
IUPAC Name:
2-(2,3-dimethylphenyl)acetic acid
Synonyms:
(2,3-dimethylphenyl)acetic acid
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
AMPNBNGNPLXMOP-UHFFFAOYSA-N
SMILES:
CC1=C(C)C(CC(O)=O)=CC=C1
1
–
1
of
1
results
1
2,3-Dimethylphenylacetic acid, 95%
CAS: 30981-98-7 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.20 MDL Number: MFCD03844736 InChI Key: AMPNBNGNPLXMOP-UHFFFAOYSA-N Synonym: 2,3-dimethylphenylacetic acid,2-2,3-dimethylphenyl acetic acid,2,3-dimethylbenzeneacetic acid,dimethylbenzeneacetic acid,2,3-dimethylphenyl acetic acid,2,3-dimethylphenylaceticacid,xylene-acetic acid,pubchem2466,acmc-1cldr,2-2,3-dimethylphenyl aceticacid PubChem CID: 20572731 IUPAC Name: 2-(2,3-dimethylphenyl)acetic acid SMILES: CC1=C(C)C(CC(O)=O)=CC=C1