accessibility menu, dialog, popup

UserName

CH3H2NO(E/Z)

CAS RN 30989-81-2

IUPAC Name: 3-amino-2-methylprop-2-enal
Synonyms: 3-amino-2-methylprop-2-enal
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: OEZAAXHZEMTBOV-UHFFFAOYSA-N
SMILES: CC(=CN)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results