accessibility menu, dialog, popup

UserName

CAS RN 31168-10-2

IUPAC Name: 3-(4-chlorophenoxy)pentane-2,4-dione
Synonyms: 3-(4-chlorophenoxy)pentane-2,4-dione
Molecular Weight (g/mol): 226.66
Molecular Formula: C11H11ClO3
InChi Key: QOOLFYCEZZCVSE-UHFFFAOYSA-N
SMILES: CC(=O)C(OC1=CC=C(Cl)C=C1)C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)