CAS RN 31405-60-4
CAS RN 31405-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5'-Hydroxy-2'-methoxyacetophenone, TRC
CAS: 31405-60-4 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.17 IUPAC Name: 1-(5-hydroxy-2-methoxyphenyl)ethanone SMILES: COc1ccc(O)cc1C(=O)C