accessibility menu, dialog, popup

UserName

NClNHSH2N

CAS RN 31430-27-0

IUPAC Name: (5-chloropyridin-2-yl)thiourea
Synonyms: 5-chloropyridin-2-ylthiourea
Molecular Weight (g/mol): 187.65
Molecular Formula: C6H6ClN3S
InChi Key: WEGKERUDVFDIST-UHFFFAOYSA-N
SMILES: NC(=S)NC1=NC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results