accessibility menu, dialog, popup

UserName

H3COOOOOHHO

CAS RN 3162-96-7

IUPAC Name: 6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-7,8-diol
Synonyms: 6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-7,8-diol
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H18O6
InChi Key: VVSWDMJYIDBTMV-UHFFFAOYNA-N
SMILES: COC1OC2COC(OC2C(O)C1O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results