accessibility menu, dialog, popup

UserName

CAS RN 3163-27-7

Br

CAS RN 3163-27-7

IUPAC Name: 1-(bromomethyl)naphthalene
Synonyms: 1-(bromomethyl)naphthalene
Molecular Weight (g/mol): 221.1
Molecular Formula: C11H9Br
InChi Key: RZJGKPNCYQZFGR-UHFFFAOYSA-N
SMILES: BrCC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results