accessibility menu, dialog, popup

UserName

CAS RN 3164-29-2

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid diamine
Synonyms: L(+)-tartaric acid diamine
Molecular Weight (g/mol): 184.15
Molecular Formula: C4H12N2O6
InChi Key: NGPGDYLVALNKEG-OLXYHTOASA-N
SMILES: N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Physical Form 
  • (3)
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)