CAS RN 31696-67-0
CAS RN 31696-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy-9H-thioxanthen-9-one, TRC
CAS: 31696-67-0 Molecular Formula: C13H8O2S Molecular Weight (g/mol): 228.27 Synonym: 2-Hydroxythioxanthen-9-one,2-Hydroxythioxanthone IUPAC Name: 2-hydroxythioxanthen-9-one SMILES: Oc1ccc2Sc3ccccc3C(=O)c2c1