accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 3188-00-9

IUPAC Name: 2-methyloxolan-3-one
Synonyms: coffee furanone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: FCWYQRVIQDNGBI-UHFFFAOYNA-N
SMILES: CC1OCCC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)