CAS RN 3188-13-4
CAS RN 3188-13-4
IUPAC Name:
(chloromethoxy)ethane
Synonyms:
chloromethyl ethyl ether
Molecular Weight (g/mol):
94.54
Molecular Formula:
C3H7ClO
InChi Key:
FCYRSDMGOLYDHL-UHFFFAOYSA-N
SMILES:
CCOCCl