accessibility menu, dialog, popup

UserName

FFFOO

CAS RN 32042-38-9

IUPAC Name: 2,2,2-trifluoroethyl formate
Synonyms: 2,2,2-trifluoroethyl formate
Molecular Weight (g/mol): 128.05
Molecular Formula: C3H3F3O2
InChi Key: CAFROQYMUICGNO-UHFFFAOYSA-N
SMILES: FC(F)(F)COC=O
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results