CAS RN 32136-81-5
CAS RN 32136-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methoxy-4'-hydroxyacetophenone, TRC
CAS: 32136-81-5 Molecular Formula: C9 H10 O3 Molecular Weight (g/mol): 166.17 Synonym: Ethanone, 1-(4-hydroxyphenyl)-2-methoxy-,Acetophenone, 4'-hydroxy-2-methoxy- (8CI),1-(4-Hydroxyphenyl)-2-methoxyethanone,1-(4-Hydroxyphenyl)-2-methoxyethan-1-one,2-Methoxy-1-(4-hydroxyphenyl)ethanone,2-Methoxy-4'-hydroxyacetophenone,4-Hydroxy-α-methoxyacetophenone IUPAC Name: 1-(4-hydroxyphenyl)-2-methoxyethanone SMILES: COCC(=O)c1ccc(O)cc1